Would you like to log in again?
VWR® Individually Calibrated Liquid-In-Glass Thermometer
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
60205-1311 602010111 602010211 602051311 60205-1111 60205-0111 60201-0111 602050311 60201-0211 60205-0011 60205-1211 60205-0311 60205-0111
89095-638EA 441.61 USD InStock
CA89095-638 CA89095-636 CA89095-668 89095-638 89095-664 89095-632 89095-630 89095-668 CA89095-634 89095-636 CA89095-632 89095-666 89095-634
VWR® Individually Calibrated Liquid-In-Glass Thermometer
Thermometers Analog Thermometers
Individually calibrated, these thermometers feature blue organic liquid fill against lead-free white back glass.
  • Individually serialized
  • Lead-free glass
Ideal for use in chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical/medical industry, manufacturing, dairies, breweries and distilleries.
Certifications: NIST.
Ordering information: Includes individual Thermometer Calibration Report stating corrections to one-tenth of the smallest division, unique report number and NIST standards used for the calibration.
Order Now
ORDER
SERVICES
Services: VWRCATALYST™ offers convenient and efficient re-calibration services for your liquid in glass thermometers with an industry leading 5-day average turn-around time from receipt of your instrument. Simply order the stated catalog no. based on how many calibration points are desired. You will receive a protective box to ship your instrument to Avantor's certified calibration laboratory along with a work order form to specify your calibration points 0 to 100°C (32 to 212°F) and prepaid return shipping label. Upon receipt at the laboratory your instrument will be inspected for any defect under a microscope and the performance of your instrument will be compared to certified standards and corrections to one-tenth of the smallest division recorded on your signed calibration report. Your calibration report and instrument are carefully repacked then returned to you prepaid.
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty