Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® PFA Safety-Coated Liquid-In-Glass Thermometers
Supplier: VWR
VWR®
607100811 60710-1011 60710-0811 60710-0211 607100211 607100611 60710-0911 60710-1411 607101011 60710-1311 60710-0311 60710-1111 60710-0611 60710-0511 60710-1211
89095-660CS 385.79 USD InStock
89095-660 89095-650 CA89095-658 89095-648 89095-653 89095-642 89095-654 89095-640 89095-646 CA89095-644 89095-658 89095-644 CA89095-654 CA89095-642 89095-656
VWR® PFA Safety-Coated Liquid-In-Glass Thermometers
Thermometers Analog Thermometers
Mercury free thermometers ideal for use in chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical/medical industry, manufacturing, dairies, breweries and distilleries.
  • PFA safety coating keeps thermometer liquid and glass encapsulated in the event of breakage
  • Contain blue organic liquid against lead-free white back glass
  • Tested and inspected by our USA quality team
  • Coated thermometers are just as accurate as uncoated thermometers, but are not recommended for timed tests as the coating can slow reaction time
Certifications: Traceable to NIST, individually serialized.
Ordering information: Include multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability.
Order Now
ORDER
SERVICES
Services: VWRCATALYST™ offers convenient and efficient re-calibration services for your liquid in glass thermometers with an industry leading 5-day average turn-around time from receipt of your instrument. Simply order the stated catalog no. based on how many calibration points are desired. You will receive a protective box to ship your instrument to Avantor's certified calibration laboratory along with a work order form to specify your calibration points 0 to 100°C (32 to 212°F) and prepaid return shipping label. Upon receipt at the laboratory your instrument will be inspected for any defect under a microscope and the performance of your instrument will be compared to certified standards and corrections to one-tenth of the smallest division recorded on your signed calibration report. Your calibration report and instrument are carefully repacked then returned to you prepaid.
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty