Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Verification Thermometers
Supplier: VWR
VWR®
606000811 60600-1111 606001011 606001311 60600-0611 60600-0811 606000611 60600-0511 606000511 60600-1611 60600-0911 60600-1311 606000911 60600-0711 60600-1011 60600-1511 606001111 606000711
89095-690CS 1557.35 USD InStock
89095-690 75831-366 CA89095-692 CA89095-670 CA89095-680 89095-680 CA89095-690 89095-670 89095-692 89095-686 89095-676 CA89095-684 89095-684 CA89095-678 75831-368 CA89095-676 CA89095-686 89095-678
VWR® Verification Thermometers
Thermometers Analog Thermometers
To verify sample temperatures in freezers, refrigerators, incubators, or ovens with traceability to NIST.
  • Accurate temperature reading without sample contamination is achieved by using the filled bottle (included), which mimics the sample
  • Insert thermometer into the bottle and place near the samples for best results
  • Thermometers contain organic liquid; feature permanently fused markings and lead free glass
  • Bottle contains glass beads or organic liquid
  • Manufactured and tested by our USA quality team; individually serialized
Statement of Accuracy indicates accuracy traceable to NIST for every thermometer.
Ordering information: Includes multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability.
Order Now
ORDER
ACCESSORIES
SERVICES
Services: VWRCATALYST™ offers convenient and efficient re-calibration services for your liquid in glass thermometers with an industry leading 5-day average turn-around time from receipt of your instrument. Simply order the stated catalog no. based on how many calibration points are desired. You will receive a protective box to ship your instrument to Avantor's certified calibration laboratory along with a work order form to specify your calibration points 0 to 100°C (32 to 212°F) and prepaid return shipping label. Upon receipt at the laboratory your instrument will be inspected for any defect under a microscope and the performance of your instrument will be compared to certified standards and corrections to one-tenth of the smallest division recorded on your signed calibration report. Your calibration report and instrument are carefully repacked then returned to you prepaid.
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty