Would you like to log in again?
VWR® Certified Precision Verification Thermometers
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
60207-0111 60207-0011 602070011 60207-0211
89095-698CS 175.33 USD InStock
CA89095-696 89095-698 89095-696 89095-700
VWR® Certified Precision Verification Thermometers
Thermometers Analog Thermometers
Used to verify sample temperatures in freezers, refrigerators, or incubators with traceability to NIST.
  • Organic liquid
  • Each item features individual serial numbers and a Statement of Accuracy indicating accuracy traceable to NIST
Tested and calibrated in an exclusive triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory against equipment whose calibration is traceable to NIST and DKD/PTB.
Each item features individual serial numbers and a Calibration Report stating corrections to one-tenth of the smallest division. Organic liquid.
Order Now
ORDER
SERVICES
Services: VWRCATALYST™ offers convenient and efficient re-calibration services for your liquid in glass thermometers with an industry leading 5-day average turn-around time from receipt of your instrument. Simply order the stated catalog no. based on how many calibration points are desired. You will receive a protective box to ship your instrument to Avantor's certified calibration laboratory along with a work order form to specify your calibration points 0 to 100°C (32 to 212°F) and prepaid return shipping label. Upon receipt at the laboratory your instrument will be inspected for any defect under a microscope and the performance of your instrument will be compared to certified standards and corrections to one-tenth of the smallest division recorded on your signed calibration report. Your calibration report and instrument are carefully repacked then returned to you prepaid.
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty