Would you like to log in again?
Nalgene® Polypropylene Economy Bottles, Wide Mouth, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
2187-0032 2187-0001 2187-0004
89096-772CS 258.78 USD InStock
89096-772 89096-762 89096-766
Nalgene® Polypropylene Economy Bottles, Wide Mouth, Thermo Scientific
Bottles Plastic Bottles
Nalgene® Economy Bottles are lightweight, more flexible versions of the original Nalgene Lab Quality Bottles for less rigorous lab use.
  • Bottles are autoclavable, guaranteed leakproof
  • made in our ISO-certified USA manufacturing facility
These Nalgene® Economy Bottles feature excellent impact and chemical resistance and clarity, include a polypropylene closure, and are made from natural, translucent polypropylene copolymer.
Priced below low-quality plastic or glass bottle brands, Nalgene® Economy Bottles are the economical, quality choice for lighter-duty lab use.
These bottles are suitable for light-duty, general purpose laboratory applications such as containing samples and daily working solutions like buffers and bench reagents.
Ordering information: Thermo Scientific Nalgene® High-Density Polyethylene Environmental Sample Bottles are also available; contact your Avantor sales representative for more information.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty