Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Round-Bottom Pressure Flasks, Ace Glass
Supplier: Ace Glass
8417-13 8417-05 8415-15 8415-21 8417-17 8417-03 8417-09 8415-25 8415-11 8417-15 8415-17 8415-07 8417-07 8415-05
89097-196EA 256.33 USD InStock
89097-196 89097-186 89097-194 89097-192 89097-190 89097-204 89097-202 89097-200 89097-212 89097-198 89097-210 89097-188 89097-208 89097-206
Round-Bottom Pressure Flasks, Ace Glass
Reaction Vessels Reaction Vessels, Flask Shape
Heavy-wall, round-bottom flasks are pressure rated at 60psi up to 120°C (248°F)
The round bottom facilitates use in heating mantles. Flasks come with a PTFE front-seal plug and FETFE O-ring for a secure seal. Flasks with a thermowell port accommodate thermometers and temperature sensors. Pressure ratings are based on ambient temperatures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty