Would you like to log in again?
Stirrer Bearing, PTFE, Ace Glass Incorporated
Supplier: Ace Glass
13443-12 13443-10 13443-08 13443-06
89082-960EA 1195.62 USD InStock
89082-960 89082-950 89097-292 89082-956
Stirrer Bearing, PTFE, Ace Glass Incorporated
Stirrers Stirring Tools
Inert PTFE stirrer bearings feature a totally enclosed bearing body for non-shedding, anti-whip, chemically resistant performance
The design has a composite PTFE/PEEK main internal seal and a specially fabricated glass ball-bearing for rigidity and smoothness during lengthy operation. The maximum recommended speed is 500 rpm in continuous operation. Ideal for glass, metal, or PTFE stir shafts. The bottom is a molded, inner standard taper joint. Average length is 96 mm, with a 45 mm O.D. at bearing center.
The bearings can also be used in slight vacuum or slight pressure applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty