Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Offset Stir Motors, Standard and Hazardous Duty, Ace Glass
Supplier: Ace Glass
13557-240 13557-220 13557-420 13557-440
89097-302EA 11029.43 USD InStock
89097-302 89097-310 89097-300 89097-308
Offset Stir Motors, Standard and Hazardous Duty, Ace Glass
Stirrers Overhead Stirrers
Offset gearbox and motors come complete with a right angle drive unit for pilot plant systems.
  • Motors available for either standard or hazardous duty applications
  • Available in either 1/4 or 1/2 hp
  • Motors have a digital 115/230 V 50/60 Hz cULus rated control
Motors with right-angle drive save headspace and work well where height is restricted.
Certifications: Hazardous duty version is UL Classified and meets Class I, Group D, T3C for flammable gases and vapor atmosphere operation and Class II, Div 1 Group F&G classification for flammable dust.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty