Would you like to log in again?
KimCote® Reservoir Bottles with Bottom Hose Outlet, DWK Life Sciences
Supplier: DWK Life Sciences
KC14607-10000 KC14607-500 KC14607-2000 KC14607-5000 KC14607-1000 KC14607-20000 KC14607-250
89097-336CS 467.37 USD InStock
89097-336 89097-334 89097-332 89097-330 89097-340 89097-338 89097-328
KimCote® Reservoir Bottles with Bottom Hose Outlet, DWK Life Sciences
Bottles Plastic Bottles
These reservoir bottles are designed to store and discharge liquids via a bottom hose outlet.
  • KimCote® plastic safety coating
  • Bottom hose glass outlet is fused to the bottle
  • Tooled top and narrow mouth
  • White enamel marking spot
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty