Would you like to log in again?
KIMBLE® KimCote® Heavy Duty Serum Bottles, DWK Life Sciences
Supplier: DWK Life Sciences
KIMBLE®
KC14960-4 KC14960-9
89097-344CS 661.62 USD InStock
89097-344 89097-342
KIMBLE® KimCote® Heavy Duty Serum Bottles, DWK Life Sciences
Bottles Glass Bottles
The heavy duty construction of these bottles is designed to prolong life expectancy with harder than normal usage. Ideal for packaging and storage.
  • Plastic-coated bottle with rounded shoulders
  • Neck is tooled for a uniform fit with a #8 rubber stopper
Manufactured from USP Type I borosilicate molded glass.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty