Would you like to log in again?
KIMAX® KimCote® Heavy-Duty Solution Bottles, DWK Life Sciences
Supplier: DWK Life Sciences
KIMAX®
KC14950-35 KC14950-25
89097-348CS 653.15 USD InStock
89097-348 89097-346
KIMAX® KimCote® Heavy-Duty Solution Bottles, DWK Life Sciences
Bottles Solution Bottles
KimCote® plastic safety coated solution bottles with carboy clamp and silicone stopper assemblies for solution bottles.
  • Neck is tooled for a #12 stopper
  • Five gallon size is designed from Federal Specification DD-B-597
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty