Would you like to log in again?
KimCote® Solution Bottles for use with Rubber Stoppers, ® Narrow Mouth, DWK Lifesciences
Supplier: DWK Life Sciences
KC15093-20000 KC15093-10000
89097-356CS 997.12 USD InStock
89097-356 89097-354
KimCote® Solution Bottles for use with Rubber Stoppers, ® Narrow Mouth, DWK Lifesciences
Bottles Solution Bottles
Narrow mouth solution bottles for use with rubber stoppers.
  • KimCote® plastic safety coating
  • Uses 11.5 stopper
Certifications: Meets USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty