Would you like to log in again?
KimCote® Solution Bottles with Color-Coded PTFE Flathead Stoppers, DWK Life Sciences
Supplier: DWK Life Sciences
KC15097-250 KC15097-500 KC15097-1000 KC15097-100 KC15097-2000
89097-358CS 2035 USD InStock
89097-358 89097-366 89097-364 89097-362 89097-360
KimCote® Solution Bottles with Color-Coded PTFE Flathead Stoppers, DWK Life Sciences
Bottles Solution Bottles
KimCote® solution bottles are designed for storage of solutions and ideal for media preparations.
  • Narrow mouth increases mechanical strength
  • Bottle necks are standard taper ground to accept flathead color-coded PTFE stoppers
  • Manufactured from 33 expansion, low extractable borosilicate glass
  • Replacement stoppers are 41941
Autoclavable safety coating reduces the hazards of shattered glass and leakage of toxic or corrosive chemicals.
Certifications: Meets USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty