Would you like to log in again?
KIMBLE® KimCote® GL 45 Media Bottles, DWK Life Sciences
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
KC14395-250 KC14395-100 KC14395-2000 KC14395-1000 KC14395-5000 KC14395-500 KC14395-10000
89097-378CS 561.27 USD InStock
89097-378 89097-368 89097-376 89097-374 89097-372 89097-370 89097-380
KIMBLE® KimCote® GL 45 Media Bottles, DWK Life Sciences
Bottles Cell Culture Bottles
Ideal for general laboratory use including mixing, storing or transporting culture media, chemicals, or solvents.
  • 30 mm Int.Ø. opening
  • Blue polypropylene linerless GL 45 screw thread cap
  • Enhanced graduations and marking spot made with chemically resistant white enamel paint
Autoclavable KimCote® safety coating reduces the hazards of shattered glass and leakage of toxic or corrosive chemicals.
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Ordering information: Blue caps and clear pour rings included.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty