Would you like to log in again?
KIMAX® KimCote® Culture Tubes, Reusable, with Unattached Caps, Kimble Chase
Supplier: DWK Life Sciences
KIMAX®
KC45066-25150 KC45066-16125 KC45066-20150 KC45066-13100
89097-446CS 492.59 USD InStock
89097-446 89097-444 89097-442 89097-440
KIMAX® KimCote® Culture Tubes, Reusable, with Unattached Caps, Kimble Chase
Tubes Culture Tubes
KimCote® plastic-coated culture tubes are made from tubing with uniform wall thickness for maximum heat transfer and chemical resistance.
  • Ideal for biohazard test protocols
  • Supplied with phenolic screw-thread caps, unattached
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
The safety coating reduces the hazards of shattered glass and leakage of toxic or hazardous materials.
Certifications: ASTM E982, Type VI, Class A requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty