Would you like to log in again?
VWR® Entrance Mats with Stabilizing Frame
Supplier: Avantor
VWR®
PF 3150 31 W01 PF 2531 31 W03 PF 2531 31 W01 PF 3150 61 W02 PF 2531 61 W01 PF 3150 61 W01 PF 3150 31 W03 PF 3150 31 W02 PF 3150 61 W03 PF 2531 61 W03 PF 2531 61 W02 PF 2531 31 W02
89097-512EA 256.24 USD InStock
89097-512 89097-502 89097-510 89097-500 89097-498 89097-496 89097-494 89097-508 89097-492 89097-490 89097-506 89097-504
VWR® Entrance Mats with Stabilizing Frame
Floor Mats Tacky Mats
Entryway contamination control systems consist of 30- or 60-layered mats applied to a reusable white polystyrene frame.
  • Nonskid backing
  • Effectively captures soil, dirt, and dust
  • Frames are reusable
  • Mats can be easily removed while cleaning
Each layer is coated with an adhesive formulated to capture soil and contain it until the layer is peeled away and discarded
Placing mats in high-traffic areas and entryways will significantly decrease the amount of dust and dirt that typically tracks in from foot traffic and equipment wheels, reduce maintenance and cleaning costs, and prolong the life of floor finishes and carpets. Mats can be easily removed during routine floor maintenance. Frames are reusable with VWR® PureStep Adhesive Mats (see 87004-016 series).
Nonskid backing applied to the bottom of the frame provides stability on carpets, tile, concrete, linoleum, wood and other floor surfaces, allowing the mat to be used on surfaces that are not ideal for tacky mats.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty