Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 25, 2024. During this time, our site will be not available. We appreciate your patience.
SP Bel-Art Snap Cap Vials, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a part of SP
SCIENCEWARE®
F17575-0005 F17575-0010 F17575-0013 F17575-0007 F17575-0030 F17575-0020 F17575-0015 F17575-0003 F17575-0040
89097-524PK 149.95 USD InStock
89097-524 89097-532 89097-522 89097-530 89097-520 89097-518 89097-516 89097-528 89097-526
SP Bel-Art Snap Cap Vials, Bel-Art Products, a part of SP
Vials
Constructed of glass-clear styrene for complete visibility, these vials are excellent for storing dry materials, powders, tablets and other samples.
  • Clear styrene construction
  • Snap cap design
  • Excellent for dry storage
  • Rigid, heavy walls
The heavy, rigid-wall construction ensures long service and can be used at temperatures from –20 to 70 °C (–4 to 158°F). Available in nine (9) sizes from 3 dram (11 ml) through 40 dram (148 ml), they are supplied with tight-fitting, polyethylene snap-on caps.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty