Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Methyl-2-pyrrolidone (NMP), Ultrapure Solutions®, Semiconductor grade

Supplier: FUJIFILM ULTRA PURE SOLUTIONS PC
Synonyms: N-Methyl-alpha-pyrrolidone1-Methyl-2-pyrrolidoneNMPN-Methyl-2-pyrrolidoneN-Methylpyrrolidone1-Methyl-2-pyrrolidinoneN-MethylpyrrolidinoneN-methylpyrrolidoneN-Methyl-gamma-butyrolactamN-Methyl-2-pyrrolidinoneM-Pyrol

28016154
89097-773CS 817 USD InStock
89097-773
N-Methyl-2-pyrrolidone (NMP), Ultrapure Solutions®, Semiconductor grade
N-Methyl-2-pyrrolidone (NMP)
FUJIFILM Ultra Pure Solutions, Inc. is one of the largest manufacturers of high purity chemicals in North America, serving the semiconductor, disk drive, flat panel, pharmaceutical, and bio-tech industries.


Formula: C₅H₉NO
MW: 99.13 g/mol
MDL Number: MFCD00003193
CAS Number: 872-50-4
https://us-legacy.vwr.com/stibo/web/std.lang.all/88/54/2298854.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty