Would you like to log in again?
VWR® Slide Labels
Supplier: VWR
VWR®
VWR-L16-C VWR-L20-C VWR-L15-C VWR-L10-C
89097-842CS 23.51 USD InStock
89097-842 89097-854 89097-832 89097-844
VWR® Slide Labels
Labels
Designed to help prevent curling or detaching, slide labels are printed on smudgeproof litho sheets and feature a permanent adhesive.
  • Labels have a thickness 0.012 cm (0.0078")
  • Color: white
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty