Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Slide Labels

Supplier: VWR

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

VWR®
VWR-L16-C VWR-L20-C VWR-L15-C VWR-L10-C
89097-842CS 23.51 USD InStock
89097-842 89097-854 89097-832 89097-844
VWR® Slide Labels
Labels
Designed to help prevent curling or detaching, slide labels are printed on smudgeproof litho sheets and feature a permanent adhesive.

  • Labels have a thickness 0.012 cm (0.0078")
  • Color: white

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty