Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-UNC45 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
SRA-1800F ADI-SRA-1800-F SRA-1800D ADI-SRA-1800-D
89106-614EA 573 USD InStock
89106-614 CA89106-612 89106-612 CA89106-614
Anti-UNC45 Mouse Monoclonal Antibody
Antibodies
GC UNC45 (general cell UNC45/unc45a) is an Hsp90 co-chaperone that has been shown to play a critical role in the chaperoning of myosin proteins as well as the progesterone receptor (PR). There are two known vertebrate isoforms of UNC45 that are functional orthologs of the C. elegans unc-45, an important protein in the development of organized body wall muscles. The vertebrate general cell form (GC UNC45/unc45a) is ubiquitously expressed, while the striated muscle-specific isoform (SM UNC45/unc45b) is found in cardiac and skeletal muscle. In vitro, GC UNC45 preferentially binds the Hsp90 beta isoform, and is specifically required for proper cellular distribution of Hsp90β but not Hsp90α.

  • For Western Blot

Recognizes human, mouse and rat UNC45. Detects a band of ~98kDa by Western blot.

Type: Primary
Antigen: UNC45
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty