Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CLIP Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
ADI-CSA-600-F ADI-CSA-600-D
89106-678EA 547 USD InStock
89106-678 89106-676
Anti-CLIP Mouse Monoclonal Antibody
Antibodies
CLIP (class II-associated invariant chain peptide) is an important peptide in the MHC-II antigen processing pathway, formed by the enzymatic cleavage of amino acids 81-104 from the invariant chain. During MHC-II synthesis in the ER and transport to endocytic vesicles, the antigen binding site of MHC-II is obstructed by the presence of the invariant chain. As the MHC-II complex moves through the Golgi, the invariant chain is degraded, leaving the CLIP fragment in the MHC-II peptide binding groove. CLIP is eventually displaced by the loading of antigenic peptides, followed by presentation on the APC cell surface to CD4+ T-cells.

  • For Immunoprecipitation

Recognizes human CLIP. Detects a band of ~33-35kDa by Western blot.

Type: Primary
Antigen: CLIP
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty