Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Cu/Zn SOD Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
ADI-SOD-100-F SOD-100J ADI-SOD-100-J ADI-SOD-100-D
89106-702EA 565 USD InStock
89106-702 89106-700 95046-094 CA95046-094
Anti-Cu/Zn SOD Rabbit Polyclonal Antibody
Antibodies
SOD (Superoxide dismutase) is responsible for the elimination of cytotoxic active oxygen by catalyzing the dismutation of the superoxide radical to oxygen and hydrogen peroxide. There are three SOD isoenzymes in mammalian cells, they are: EC SOD (extracellular SOD), Cu/Zn SOD (copper and zinc-containing SOD) and Mn SOD (manganese-containing SOD).

  • For Immunofluorescence, Immunoprecipitation, Western Blot

Recognizes human, mouse, rat, and clam Cu/Zn SOD. Detects a band of ~23kDa (human) or ~19kDa (mouse and rat) by Western blot.

Type: Primary
Antigen: Cu/Zn SOD
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty