Would you like to log in again?
SP Bel-Art ProCulture Petri Dish Rack, 60 mm, Bel-Art Products, a part of SP
Supplier: Bel-Art
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SCIENCEWARE®
F18991-0060
89106-748EA 112.59 USD InStock
89106-748
SP Bel-Art ProCulture Petri Dish Rack, 60 mm, Bel-Art Products, a part of SP
Racks Petri Dish Racks
This rack is useful for safe transport and incubation of 60 mm Petri dishes and helps to streamline benchtop sample processing.
  • Small benchtop footprint
  • Stackable
  • Finger access slot allow safe removal from the rack with a secure grip
  • Autoclavable
Each rack has six columns and holds up to fifty-four Petri dishes with lids
Up to four racks can be stacked together and interlocked to prevent accidental knock over. Finger access slots in the base allow a stack of dishes to be safely removed from the rack with a secure grip. Rack base is epoxy coated steel, and all other parts are made of white polypropylene.
Steam autoclavable at 121 °C (250 °F). Racks are shipped flat and easily assemble in minutes.
The 101/2×63/4" base area preserves benchtop space, and a large center divider has space for labeling and includes a carrying handle that does not interfere with samples.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty