Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Nunc® IVF ICSI Dish, Thermo Scientific
Supplier: Thermo Scientific
Nunc®
150265 150265
89106-782CS 178.12 USD InStock
89106-782 CA89106-782
Nunc® IVF ICSI Dish, Thermo Scientific
Culture Dishes Petri Dishes
Thermo Scientific Nunc® provide an extensive portfolio of products intended for use in human In Vitro Fertilization (IVF)
The Nunc® IVF ICSI Dish maintains an optimal temperature with true flat bottom, as well as increased safety with the easy grip design. The low profile provides optimal angles during ICSI process, while the super polished optical center allows clear viewing of the sample.
Thermo Scientific Nunc® InVitro Fertilization Products provide security, safety and reproducibility for your valuable samples, and are uniquely certified based on a 1-cell stage mouse embryo toxicity test. Stringent lot-level manufacturing controls ensure viability, reliability and reproducibility. Lid is Present.
Products are produced under ISO 13485, MEA tested, CE marked class II medical device and FDA cleared for use in IVF.
Certifications: CE and FDA
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty