Would you like to log in again?
IMPINGER SET- COMPLETE
Supplier: GENERAL GLASS BLOWING CO
Impinger^ Also called a bubbler, is a two piece set consisting of: a) impinger bottom with an etch mark at the 12ml b) impinger top with a restricted tip that fits into the bottom. the ground glass connection makes a tight, leakproof seal at the neck level
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
IMPINGER SET- COMPLETE 89107-074 GW200 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty