Would you like to log in again?
NorthFlex™ Cold Grip Winter Gloves, Honeywell Safety
Supplier: Honeywell Safety
NorthFlex™
NF11HD/8M NF11HD/9L
89107-118CS 1130.02 USD InStock
89107-118 89107-120
NorthFlex™ Cold Grip Winter Gloves, Honeywell Safety
Gloves
These gloves are ideal for handling damp, oily, or greasy parts in cold conditions.
  • Polyamide knitted with PVC foam coating
They are well suited for many tasks where workers need protection from the cold.
Certifications: Certified to XX EN 420, EN 388:2231, EN 511:X2X.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty