Would you like to log in again?
WorkEasy® Polyurethane-Coated Industrial Gloves, Honeywell Safety
Supplier: Honeywell Safety
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
WE300-M WE300-XL WE300-S WE300-L
89107-227PK 74.21 USD InStock
89107-227 89107-229 89107-223 89107-225
WorkEasy® Polyurethane-Coated Industrial Gloves, Honeywell Safety
Gloves
The WorkEasy® WE300 glove series features a new and improved fiber blend that provides a higher level of cut and abrasion protection.
  • Dyneema®, fiberglass, and polyester blend knit shell provides outstanding cut and slash protection
  • Polyurethane coating adds abrasion, tear, and puncture resistance
  • Lightweight for excellent dexterity
  • Gloves are easily laundered for longer life
These lightweight yet durable industrial gloves ensure reliable use in small parts assembly, fabrication, inspections, electronic assembly, automotive sub-assembly, parts handling, recycling/glass handling, shipping/receiving, and warehousing applications.
Workers appreciate the bare-handed feel of the lightweight knit shell for excellent dexterity and tactile sensitivity. The technologically improved fiber blend offers a consistantly high level of mechanical cut protection for applications where cut or slash hazards are a high risk.
Certifications: ANSI Cut Level 3. EN 388 Cut Level 5, Abrasion Level 4.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty