Would you like to log in again?
Bench Top Reactor Stand, Ace Glass Incorporated
Supplier: Ace Glass
12840-10
89107-394EA 6563.42 USD InStock
89107-394
Bench Top Reactor Stand, Ace Glass Incorporated
Support Stands
This newer bench reactor stand assembly accommodates all types of glass bench filter and pressure reactors.
The stand is compatible with reactors from 1 to 6 Liters having overhead drive motors, and in both jacketed and non-jacketed flask versions. The bottom plate, motor mount and middle flask supports are all adjustable to fit any small reactor, and the stand will fit into most laboratory hoods. It comes completely assembled, but it is very easy to take apart for cleaning.
The open bottom accommodates round-bottom and flat-bottom flasks, and even flasks with bottom valves. Will accept either IKA or Heidolph overhead stir motors. All stand components are either anodized or powder coated aluminum. The flasks sit on an attached rubber base plate.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty