Would you like to log in again?
Tuff Coat™ Cut Resistant Gloves, Honeywell Safety
Supplier: Honeywell Safety
KV200-XL KV200-L
89107-938CS 2360.34 USD InStock
89107-938 89107-940
Tuff Coat™ Cut Resistant Gloves, Honeywell Safety
Gloves
Tuff-Coat liners feature 10-cut heavy weight DuPont® KEVLAR™ brand fiber that are soft and comfortable, with no seams to rub and chafe.
  • Cut-resistant seamless knit liner
  • Natural rubber palm and fingertip coating
  • Rubber coating provides excellent grip and flexibility
  • Durable hand protection without sacrificing dexterity
  • Excellent protection against cuts and abrasions
Ideal for use in construction, forestry, municipal services and transportation.
Certifications: ANSI/ASTM rated cut-level 2.
Caution: Gloves are not cut or puncture proof, not recommended for use with moving or serrated blades.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty