Would you like to log in again?
QuickConnect™ Mop Handles
Supplier: Contec
QuickConnect™
2645 2643 CFH114 2642 CFH70 2686 2644 CFH334 CFH224 2725E
33502-716EA 314.19 USD InStock
33502-716 33502-717 89107-996 77322-288 33502-714 33502-715 77322-290 77322-294 89201-652 77322-296
QuickConnect™ Mop Handles
Mops
Interchangeable mop handles are available in carbon fiber, anodized aluminum, stainless steel or fiberglass.
  • Compatible with all QuickConnect™ frames
  • Multiple lengths available
Fully autoclavable QuickConnect™ mop handles and interchangeable mop head frames work together for maximum flexibility when cleaning in critical environments. Invest in one handle and the various interchangeable heads required for specific applications. Handles are available in several lengths and styles.
Telescoping handles are available in lightweight carbon fiber, anodized aluminum, durable stainless steel, or fiberglass. Choose from a variety of lengths. For hard-to-reach ceilings and walls, a carbon fiber handle is available that is lightweight and easy to maneuver. A one-piece stainless-steel handle is also available.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty