Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Sterile 70% Isopropanol

Supplier: VWR
VWR®
89108-160 89499-420 89108-162 89499-420 89108-160 89108-162
89499-420CS 272 USD InStock
89499-420 CA89499-420 89108-162 CA89108-160 89108-160 CA89108-162
VWR® Sterile 70% Isopropanol
Cleaning Agents Disinfectants
Filtered to 0.2 µm and induction sealed to assure sterility.

  • Contains 70% USP grade isopropanol and 30% USP grade deionized water
  • Double-bagged and gamma-irradiated to a Sterility Assurance Level of 10-6
  • Ideal for use with sterile cleanroom wipers

Excellent for wipedown of isolators, cabinets, laminar flow hoods.

Each bottle is lot coded and has an expiration date.

Suitable for wipedown of products and equipment for pass through to controlled environments. Can be used on gloved hands in sterile environments.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty