Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 

Ethanol ≥95%, KOPTEC, meets analytical specification of BP, Ph. Eur., USP (190 Proof) (not denatured)

Supplier: Decon Labs
Danger
Synonyms: Ethyl alcohol

V1105HC V1155 V1101TP V1116 V1105SG V1116GTP V1105TP V1105HCTP V1116TP V1155MTP V1105MTP V1105 V1105M V1105SGTP V1101GTP V1101G V1155M V1101 V1155TP
89125-178CS 761.91 USD InStock
89125-178 89125-168 71002-510 89125-166 89125-164 71001-868 89234-858 71001-626 89125-162 89125-184 89125-182 89234-854 89125-180 89501-408 89234-856 71002-428 71001-630 89501-406 71002-424
Ethanol ≥95%, KOPTEC, meets analytical specification of BP, Ph. Eur., USP (190 Proof) (not denatured)
Ethanol
190 proof (95%) pure (undenatured) ethanol. Meets USP and multi-compendial specifications. Used for research, healthcare, industrial, and production applications as a cleaner, solvent, and reagent.

  • Water 5%
  • Ethyl alcohol: 95%
  • Denaturant: none


Formula: H₃CCH₂OH
MW: 46.07 g/mol
Melting Pt: –114 °C
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3,II
Order Now
Specification Test Results
Assay 190 proof 94.9-96.0 (% v/v) Ethanol
Water 4.0-5.1 (% v/v)
Specific Gravity @15.56C 0.8120-0.8160 (USP)
Specific Gravity @15.0C 0.8140-0.8160 (JP)
Relative Density @ 20C 0.8051-0.8124 (BP/EP)
Color Passes USP test for color
Clarity Passes USP Test for Clarity
Acidity Max. 30 ppm
Alkalinity Passes USP Test
Non Volatile Residue Max. 25 ppm
Sum of Acetal & Acetaldehyde compounds Max.10 ppm
Benzene Max.2 ppm
Methanol Max.100 ppm
UV Absorbance @ 240 nm Max. 0.40 AU (USP/BP/EP/JP)
UV Absorbance @ 250 nm Max. 0.30 AU (USP/BP/EP/JP)
UV Absorbance @ 260 nm Max. 0.30 AU (USP/BP/EP/JP)
UV Absorbance @ 270 nm Max. 0.10 AU (USP/BP/EP/JP)
UV Absorbance @ 235-340 nm (Smooth curve) Pass (USP/BP/EP/JP)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty