Would you like to log in again?
Wide Mouth Glass Jugs, Clear, C&G Containers
Supplier: C&G Containers
LFW320900000 LFW128900000 LFW320200010 LFW064200000 LFW064900000 LFW128200010 LFW064200010 LFW128100000
30621-208CS 79.06 USD InStock
30621-208 71000-871 89125-410 71000-873 71000-862 71000-854 71000-866 71000-858
Wide Mouth Glass Jugs, Clear, C&G Containers
Bottles Carboys
These glass jugs are used primarily for the collection of benthic organisms and the collection of water samples for mulitple analyte testing.
Pre-cleaned and certified products contain the Certificate of Analysis and Cleaning Protocol Form in the product case.
A lot certificate of analysis is generated with the analytical results obtained, including the concentration reporting limits and units of measurement, for Pre-cleaned and Certified products. This certificate of analysis is included in each product case.
Wash protocols for these jugs include; A1 (nitric acid wash), B1 (deionized water wash), C1 (hydrochloric acid wash), and L3 (no wash).
Secondary uses include containment and archiving. Please refer to the applicable Certificate of Analysis for certified analytes and corresponding reporting limits.
Ordering information: Container measurements and capacity 'contain' volumes may vary.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty