Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

WHEATON® CryoELITE® Cryogenic Vials, Freestanding, DWK Life Sciences

Supplier: DWK Life Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

CryoELITE®
W985853 W985916 W985869 W985923 W985867 W985866 W985922 W985925 W985903 W985868 W985917 W985870 W985864 W985919 W985871 W985918 W985924 W985862 W985874 W985920 W985921 W985863 W985902 W985865 W985872 W985915 W985852
89125-504CS 515.78 USD InStock
89125-504 89125-502 89125-468 89125-500 89125-466 89125-464 89125-484 30620-972 89125-482 89125-480 89125-518 89125-516 89125-514 89125-512 89125-478 89125-510 89125-498 89125-476 89125-452 89125-474 89125-472 89125-450 89125-492 89125-470 89125-490 89125-508 89125-506
WHEATON® CryoELITE® Cryogenic Vials, Freestanding, DWK Life Sciences
Vials
CryoELITE® cryogenic freestanding Vials have a Loctagon™ Vial Skirt that provides stability in the freestanding position and locks into the CryoELITE® Benchmate rack in order to provide easy open and close wiht one hand.

  • CryoELITE® Benchmate rack can be used for both freestanding and round bottom vials
  • Manufactured from polypropylene
  • Easily celaned in an automatic washer or autoclavable at 121° for 20 minutes

The patented unrivaled cap seal exceeds DOT and IATA regulations, ensuring the ultimate level of protection for samples during transportation and demanding free-thaw handling. The cryogenic vials are made from low binding, cryogenic grade virgin polypropylene.

The availability of colored caps allows for the color coding of projects along with Wheaton colored freezer and storage boxes.

The 2D Data Matrix Bar Code Bottom Inserts are for use with CryoELITE® Freestanding Cryogenic Vials. The 2D insert allows for immediate bar coding of samples. The insert can also be applied to the vial at a future date, which eliminates jeopardizing the integrity of your sample by transferring it to another vial. The 2D Data Matrix Bar Code Insert fits and locks into place in the bottom of the vial. The insert is made of the same polypropylene as the vials themselves and is laser-etched for durability.

The Loctagon™ bottom allows for the one hand removal of freestanding vial caps. Holds up to 50 cryogenic vials. Non-skid feet offer additional stability for bench work. Each well has an Int.Ø of 12.5 mm and the rack has an overall footprint of 190×100×22 mm.

The bottom format allows the optional insertion of a Wheaton 2D Data Matrix Bar Code Insert, providing unique indentifiers for traceability.

Certifications: Each lot is certified RNase/DNase and endotoxin free and non-pyrogenic, providing assurance for sample integrity.

Ordering information: Vials are available with either a patented 2' angle plug seal for external thread vials or with an internal thread. Both external and internal CryoELITE® closure formats offer unique stop-rims to provide additional security in knowing the cap is securely closed.

Order Now
 

ORDER

ACCESSORIES

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty