Would you like to log in again?
Agarose, Low Melting Point, IBI Scientific
Supplier: IBI Scientific
IB70051 IB70056 IB70050 IB70058 IB70057 IB70059
89125-536EA 988.71 USD InStock
89125-536 89125-534 89125-532 IB70050 89133-106 89133-104
Agarose, Low Melting Point, IBI Scientific
Electrophoresis Gels Agarose and Acrylamide
Low Melting Point Agarose is commonly used as a gel matrix when fragment recovery is desired after electrophoresis
Some features of IBI Low Melt Agarose when compared to standard agarose are higher clarity, greater sieving capacity, and lower gel strength yet still easy to handle.
IBI Low Melt Agarose can be used for DNA electrophoresis, RNA electrophoresis, cloning of tissue culture cells, and viral plague assays. Low Melt Agarose gels will separate and finely resolve molecules that are > 1000bp.
IBI Low Melt Agarose is derived by organic synthesis, which generates methoxylate groups from the basic agarose structure. This produces low melting and gelling temperatures when compared to standard agarose. The lower gelling temperature ensures the agarose will be in a liquid state at a temperature range where "in-gel" manipulations can be performed without prior extraction of the DNA or RNA from the gel slice. Also, the properties of low melt agarose promote quick and thorough digestion of gel slices during extraction procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty