Would you like to log in again?
Loupe Kits, Integra™ Miltex®
Supplier: Integra LifeSciences
Miltex®
1-5010 1-5004 1-5003 1-5013 1-5018 1-5006 1-5012 1-5002 1-5008 1-5019 1-5005 1-5016 1-5015 1-5017 1-5007 1-5020 1-5000 1-5001 1-5011 1-5009 1-5014
89125-812EA 2327.16 USD InStock
89125-812 89125-800 89125-822 89125-832 89125-810 89125-798 89125-820 89125-830 89125-796 89125-794 89125-792 89125-808 89125-818 89125-828 89125-806 89125-816 89125-804 89125-826 89125-814 89125-802 89125-824
Loupe Kits, Integra™ Miltex®
Magnifiers
Feature adjustable interpupillary distance, convergence, declination angle, and locking knob
Personal setting can be locked in quickly and easily. Flip up easily for quick transition to regular vision. Dual hinge design.
Ordering information: Kits include magnification loupes and frame, storage case, flip paddle, lens covers, cleaning solution, screwdriver, cleaning cloth, and head strap. An LED light source is available separately (89177-040).
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty