Would you like to log in again?
AlphaGuard® Scrub Shirts and Pants, Alpha Protech
Supplier: Alpha Protech
AlphaGuard®
WP-23322-4 WP-23322-5 WP-23322-3 WP-23322-2
89126-226CS 158.16 USD InStock
89126-226 89126-228 89126-222 89126-224
AlphaGuard® Scrub Shirts and Pants, Alpha Protech
Disposable Garments
Scrubs provide a discreet barrier for employees and are highly breathable to keep users cool and dry.
  • Safety stitched seams for added strength and durability
AlphaGuard material is extremely soft and comfortable to wear and delivers high filtration efficiency while maintaining a low particle count. Scrub shirts are available in long sleeve style with a round banded neck, one pocket, and elastic wrist. Scrub pants include a spandex waist, a rear pocket, and are reinforced along the inner thigh area.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty