Would you like to log in again?
VWR® TraceClean® Straight Side Glass Jars, Clear
Supplier: VWR
VWR®
QLFS15900241 QLAS15900269 QLFS15900247 QLFS15900259 QLFS89080078 QLFS15900255 QLFS15900261 QLFS15900245 QLFS15900243
89126-578CS 158.89 USD InStock
89126-578 89126-568 89126-596 89126-584 89126-592 89126-570 89126-580 89126-582 89094-235
VWR® TraceClean® Straight Side Glass Jars, Clear
Jars General Use Jars
These TraceClean® glass jars are used primarily for the collection of soil, sediments, and slurry media for testing Semi-volatiles, Pesticides, PCBs, Diesel Range Organics, TPH, Metals, and Phenolics.
  • Clear glass fabrication
  • Straight-sided design
  • Used as a containment vessel
  • Suited to the collection of inorganic samples
Secondary uses include containment, bench vessels, and archiving
The 950 mL clear jar 89094-235 contains 5 mL (1:1) of the chemical preservative H2SO4.
Quality Assured® with VWR® Logo Sample ID Label products, a lot certificate of analysis is generated with the analytical results obtained including the concentration reporting limits and units of measurement. This certificate of analysis is included in each product case.
Please refer to the applicable Certificate of Analysis for certified analytes and corresponding reporting limits.
Certifications: QAL.
Ordering information: Container measurements and capacity "contain" volumes may vary.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty