Would you like to log in again?
Spill-X® Spill Treatment Kit, Caustics, New Pig
Supplier: New Pig
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
GEN529
89126-674CS 520.61 USD InStock
89126-674
Spill-X® Spill Treatment Kit, Caustics, New Pig
Spill Control Materials
Packed with spill control agents that neutralize and solidify caustic spills
Works on many caustics for safe and easy cleanup, including sodium hydroxide (50%) and ammonium hydroxide (29%). Granular base lets you use only what you need; reduces product waste and disposal costs. Speeds spill cleanup to limit worker exposure and minimize production downtime. Non-toxic, free-flowing dry chemical is environmentally safe. Agents packed in easy-to-use shaker bottles. Excellent as refill for Spill-X Portable Spill Treatment Kit.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty