Would you like to log in again?
Neptune® Microcentrifuge Tubes with Attached Flat Caps, Biotix
Supplier: Biotix
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
63300631 63300646 63300637 63300629 63300640 63300645 63300641 63300642 3745.AS.X 63300643 63300635 63300632 63300638
89126-718CS 382.78 USD InStock
89126-718 89126-728 89126-714 89126-724 89126-716 89126-726 89126-710 89126-720 89126-722 89126-730 89134-504 89134-506 89134-500
Neptune® Microcentrifuge Tubes with Attached Flat Caps, Biotix
Tubes Microcentrifuge Tubes
Tubes are designed for general laboratory use and storage of aqueous solutions.
Exceptional optical clarity and well distinguished graduation marks allow easy visual reference of sample. Will withstand autoclaving at 121 °C (250 °F) at 15 psi for 15 min. Rated to RCF of 20000×g for 15 min. Tubes are frosted on the top and sides for convenient sample labeling.
Certifications: CE certified. IVD labeled. Certified free of DNA, DNase, RNase, and endotoxins (pyrogens). Manufactured under stringent controls in an ISO 9001 certified facility. Every lot is tested for precision assuring maximum quality and compatibility. All raw materials are traceable.
Ordering information: Assorted color packs include blue, green, yellow, orange, red, and lavender.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty