Would you like to log in again?
Disposable Plastic Serological Pipettes, Argos Technologies
Supplier: Argos Technologies
13002-19 13002-15 04395-22 04395-14 13002-17 13002-20 13002-29 13002-29 13002-12 13002-28 04395-13 04395-10 13002-26 13002-36 04395-20 04395-08 13002-28 04395-21 13002-24 04395-18 04395-15 13002-30 13002-37 04395-17 04395-09 13002-32 13002-14 04395-19 13002-16 04395-16
10124-806CS 431.47 USD InStock
CA11010-166 10124-806 10124-828 10124-804 10124-826 10662-288 10124-824 CA11010-164 10124-846 89126-848 75805-148 10124-792 89126-852 89126-854 89236-678 89126-850 10124-832 10124-810 10124-830 10124-838 10124-836 10124-834 89126-856 89126-858 10124-822 10124-800 10124-844 10124-842 10124-820 10124-840
Disposable Plastic Serological Pipettes, Argos Technologies
Pipettes Serological Pipettes
Argos brand standard serological pipettes are designed for accurate and rapid fluid dispensing
Argos Aspirating pipettes are sterile, individually wrapped plastic serological pipettes that are supplied without filter plugs and graduations for vacuum aspirating applications. They are very useful in tissue culture applications when aspirating larger volumes from dishes, flasks, or roller bottles.
Argos open end pipettes are just like the standard individually wrapped pipettes without the tip. The wide end opening is ideal when pipetting thick or viscous liguids, liquids that contain particulates which could block a standard tip pipette, or in applications where speed is critical. Supplied individually wrapped, graduated, sterile, and with a filter plug mouthpiece.
Argos pipettes are made from transparent medical grade polystyrene sterilized by gamma irradiation. Each lot is tested for endotoxins (C.5 Eu/ml) (ISO T0993-11), cytotoxicity (ISO10993-5), and are guaranteed non-hemolytic (ISO10993-4).
These polystyrene pipettes are sterile, pyrogen-free, and they utilize a unique filter plug to prevent overfilling. The filter plug eliminates lint fibers and is ensured to stay in place. The sharp, black, highly legible permanent graduations prevent ambiguity and guarantee precise reading and accurate dispensing to +1 to 2%. Reverse graduations show volume removed or remaining on all pipettes larger than 1 ml. Generous negative graduations provide additional pipetting volume. Pipettes are available bulk packed in polyethylene bags for high-volume usage or individually wrapped in a fiber-free paper and plastic combination that is easy to open. The uniform mouth piece fits all major pipettors. Pipettes have color-coded TD rings for ease of identification. The 1.1 ml and 2.2 ml bacteriological pipettes meet the A.P.H.A. standards for the examination of dairy products. Calibrated TD with blow out. Meets AST\< E934.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty