Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
API® M Medium Microbial Identification Kit, bioMerieux
Supplier: bioMérieux
50120 50120
89126-946EA 72.07 USD InStock
CA-89126-946 89126-946
API® M Medium Microbial Identification Kit, bioMerieux
Media for Microbiology Prepared Media for Microbiology
Tests for motility of facultative anaerobic bacteria.
  • Economical
  • Easy to use
Motility medium reagents for use with API® 20 strips.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty