Would you like to log in again?
Workstation Organizers, Menda
Supplier: Menda
35874 35873
89126-972EA 49.06 USD InStock
89126-972 89126-968
Workstation Organizers, Menda
Trays/Dishes/Pots Drawer Organization Trays
Menda workstation organizers are ideal for improving the appearance and standardizing the placement of tools at any workstation.
  • Superior strength and durability
  • Made from extruded, high-impact PP materials
  • Chemical and moisture resistant
  • Economical alternative to injection molded products
Made from an extruded conductive PP Plastek™ material, making it ideal to properly set up and standardize any ESD workstation. Conductive 10² to 10⁴ ohm material provides reliable path-to-ground.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty