Would you like to log in again?
ESD Badge Holders, Menda
Supplier: Menda
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
35013 35010 35012 35014 35015 35011
89126-984EA 4.93 USD InStock
89126-984 89126-976 89126-986 89126-980 89126-982 89126-978
ESD Badge Holders, Menda
Personal Identification ID Card Holders
ESD badge holders are ideal for use by electronic manufacturers working with ESD sensitive components.
  • Highly visible yellow header and ESD symbol allows immediate identification of ESD protection
  • Tribocharging: Less than 100 volts
  • Available in horizontal and vertical formats
Badge holders feature an inner anti-print, non-ESD vinyl liner which will prevent ink transfer from the document to the document holder. Manufactured with a custom dissipative material made exclusively for Desco Industries and not available anywhere else.
Certifications: Dissipative Rtt: 10⁶ to 10¹⁰ ohms per ANSI/ESD S11.11
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty