Would you like to log in again?
DuPont™ Tyvek® IsoClean® Frocks with Raglan Sleeves
Supplier: DuPont
Tyvek® IsoClean®
IC264SWHMD00300B IC264SWHLG00300B IC264SWHXL0030CS IC264SWHXL00300C IC264SWHLG0030CS IC264SWHLG00300C IC264SWHSM00300C IC264SWHMD0030CS IC264SWHSM00300B IC264SWHXL00300B IC264SWH3X00300B IC264SWH3X0030CS IC264SWH2X00300C IC264SWH3X00300C IC264SWHMD00300C IC264SWH4X0030CS IC264SWHSM0030CS IC264SWH4X00300C IC264SWH2X00300B IC264SWH2X0030CS IC264SWH4X00300B
89125-624CS 708.31 USD InStock
89125-624 89127-310 89127-314 10693-072 10693-070 10129-306 10129-308 10129-302 10129-444 10129-304 10016-076 10129-300 10016-078 10129-442 10016-080 10693-076 89127-306 10016-082 10016-084 10016-074 89125-626
DuPont™ Tyvek® IsoClean® Frocks with Raglan Sleeves
Disposable Garments
Tyvek® IsoClean® delivers an ideal balance of protection, durability and comfort.
  • Serged seams
  • Bound neck
  • Raglan sleeve design
  • Elastic wrists
  • Zipper closure
Made using a patented flash spinning process, Tyvek® provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam. Elastic openings at wrists and high mandarin collar with adjustable snaps allow for a secure fit. Bound neck allows for lower particle shedding. Front zipper closure allows for easy donning and doffing. Raglan sleeve design and A-line silhouette allows for greater range of motion and comfort.
Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio.
Certifications: Sterile garments are gamma sterilized to SAL of 10-6.
Packaging: Bulk packaged garments are bulk packaged in double transparent poly liners. Clean Processed and Clean Processed, Sterile garments are individually packaged.
Order Now
ORDER
SPECIFICATIONS
COMPLEMENTARY PRODUCTS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty