Would you like to log in again?
DuPont™ Tyvek® IsoClean® Shoe Covers with PVC Sole
Supplier: DuPont
Tyvek® IsoClean®
IC461SWHXL03000B IC461SWHMD03000B IC461SWHMD03000B IC461SWHLG03000B IC461SWHLG03000B IC461SWHSM03000B IC461SWHSM03000B IC461SWHXL03000B
89127-344CS 696.99 USD InStock
89127-344 CA89125-644 89125-644 CA89127-346 89127-348 CA89127-344 89127-346 CA89127-348
DuPont™ Tyvek® IsoClean® Shoe Covers with PVC Sole
Disposable Garments
Tyvek® IsoClean® delivers an ideal balance of protection, durability and comfort.
  • PVC sole
  • Elastic opening
  • Serged seams
Made using a flash spinning process, Tyvek® provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash.
Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam. Elastic at ankles allows for a secure fit. PVC soles provide enhanced skid-resistance and durability.
Packaging: Bulk packaged garments are bulk packaged in double transparent poly liners.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty