Would you like to log in again?
DuPont™ Tyvek® IsoClean® Sleeves
Supplier: DuPont
Tyvek® IsoClean®
IC501BWH0001000C IC501BWH0001000B IC501BWH0001000C IC501BWH0001000S IC501BWH000100CS IC501BWH0001000B IC501BWH0001000S IC501BWH000100CS
89127-354CS 660.66 USD InStock
CA89127-352 89127-354 89125-646 89127-352 CA89127-350 CA89127-354 89127-350 CA89125-646
DuPont™ Tyvek® IsoClean® Sleeves
Disposable Garments
Tyvek® IsoClean® delivers an ideal balance of protection, durability and comfort.
  • Elastic at both ends
  • One size fits most
  • Serged seams
Made using a patented flash spinning process, Tyvek® provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam. Elastic at both ends allows for a secure fit.
Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio.
Certifications: Sterile garments are gamma sterilized to SAL of 10-6.
Packaging: Bulk packaged option packed in double transparent poly liners.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty