Would you like to log in again?
DuPont™ Tyvek® IsoClean® Bouffant Caps
Supplier: DuPont
Tyvek® IsoClean®
IC729SWH0002500B IC729SWH0002500B IC729SWH000250CS IC729SWH000250CS IC729SWH0002500C
89127-358CS 511.27 USD InStock
CA89125-652 89127-358 89125-652 CA89127-360 89127-360
DuPont™ Tyvek® IsoClean® Bouffant Caps
Disposable Garments
Tyvek® IsoClean® delivers an ideal balance of protection, durability and comfort.
  • Serged seams
  • Elastic headband
  • One size fits most
Made using a patented flash spinning process, Tyvek® provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Serged seams have multiple interlocking threads that are sewn around the raw edges of the garment material to create a strong, stress-resistance seam. Diameter of head opening is 211/2" stretched and 6" relaxed. Elastic headband allows for one size to fit most.
Clean-processed garments are processed to minimize particle shedding.
Certifications: Sterile garments are gamma sterilized to SAL of 10-6.
Packaging: Bulk packaged option packed in double transparent poly liners.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty