Would you like to log in again?
Sampling System, Sterile, Advanced Scientifics
Supplier: Advanced Scientifics
4MP0037 4MP0038 4MP0035 4MP0039 4MP0036 4MP0034
89127-376CS 1663.25 USD InStock
89127-376 89127-374 89127-378 89127-380 89127-372 89127-382
Sampling System, Sterile, Advanced Scientifics
Bags Sample Bags
Polyethylene sample bags complete with 30.5 cm (12") c-flex tubing, pinch and cut devices, male luer injection sites, and 3.8 cm (1½") sanitary sampling system valve.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty